Merck
  • Formation of Tertiary Alcohols from the Rhodium-Catalyzed Reactions of Donor/Acceptor Carbenes with Esters.
Formation of Tertiary Alcohols from the Rhodium-Catalyzed Reactions of Donor/Acceptor Carbenes with Esters.
Organic letters (2018-04-12)
Liangbing Fu, Kevin Hoang, Cecilia Tortoreto, Wenbin Liu, Huw M L Davies
ZUSAMMENFASSUNG
Rhodium(II)-catalyzed reactions between isopropyl acetate and trichloroethyl aryldiazoacetates result in the formation of oxirane intermediates that ring open under the reaction conditions to form tertiary alcohols. When the reaction is catalyzed by the dirhodium tetrakis(triarylcyclopropanecarboxylate) complex, Rh2( S-2-Cl,4-BrTPCP)4, the tertiary alcohols are formed with good asymmetric induction (80-88% ee).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rh2(S-TCPTAD)4