Merck
A versatile, modular synthesis of monofunctionalized BODIPY dyes.
Chemical communications (Cambridge, England) (2009-07-21)
Volker Leen, Els Braeken, Kristof Luckermans, Carine Jackers, Mark Van der Auweraer, Noël Boens, Wim Dehaen
ZUSAMMENFASSUNG
A careful choice of the pyrrole building blocks allows the synthesis of a wide range of monohalogenated BODIPY dyes with excellent reactivity in palladium catalyzed coupling reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,5,6,7-Tetrahydroindol, 98%