Merck
Z692794
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT254/312)
AC/DC input 230 V AC
Größe auswählen
1 EA
€ 6.200,00
€ 6.200,00
Voraussichtliches Versanddatum23. April 2025
Größe auswählen
Ansicht ändern
1 EA
€ 6.200,00
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.22
€ 6.200,00
Voraussichtliches Versanddatum23. April 2025
Leistungsmerkmale
Contains UK and Schuko plugs
Hersteller/Markenname
Cleaver CSI-MDOCUV254/312
Allgemeine Beschreibung
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254/312 nm dual transilluminator
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254/312 nm dual tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Rechtliche Hinweise
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)