Merck
MTCP52001M
MTC Bio ProPette LE Adjustable Pipettor
capacity 100-1000 μL
Synonym(e):
pipette
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC Bio P5200-1M
Kapazität
100-1000 μL
Allgemeine Beschreibung
ProPette LE, 100 to 1000µL
Leistungsmerkmale und Vorteile
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (0.1µL-10mL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Rechtliche Hinweise
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio