Skip to Content
MilliporeSigma
Skip To
HS15906
Spatula with Flat End & Spoon End
stainless steel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Estimated to ship TODAYfromMILWAUKEE
$10.60
About This Item
NACRES:
NB.71
UNSPSC Code:
41122403
$10.60
Estimated to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
stainless steel
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS15906
L × W
150 mm × 3.8 mm
General description
Spatula transfers, lifts, and handles powders and granular materials.
Features and Benefits
  • Made from nickel-plated, stainless steel wire
  • Glazed finish with a Hayman-type end (flat and spoon)
  • Autoclavable
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
HS15907BAH367190000BAH367180000
manufacturer/tradename
Heathrow Scientific HS15906
manufacturer/tradename
Heathrow Scientific HS15907
manufacturer/tradename
Bel-Art H36719-0000
manufacturer/tradename
Bel-Art H36718-0000
material
stainless steel
material
stainless steel
material
brown FEP blade
material
stainless steel blade
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
L × W
150 mm × 3.8 mm
L × W
195 mm × 7.9 mm
L × W
-
L × W
-
sterility
non-sterile
sterility
non-sterile
sterility
-
sterility
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library