Fortfahren mit
G5755
Gly-Gly-Gly-Gly-Gly
Synonym(e):
Glycyl-glycyl-glycyl-glycyl-glycin, Pentaglycin
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
Lineare Formel:
NH2CH2CO(NHCH2CO)3NHCH2COOH
CAS-Nummer:
Molekulargewicht:
303.27
UNSPSC Code:
12352200
NACRES:
NA.26
PubChem Substance ID:
EC Number:
230-398-3
Beilstein/REAXYS Number:
1716946
MDL number:
assay
≥98% (TLC)
form
solid
color
white
application(s)
peptide synthesis
storage temp.
−20°C
SMILES string
NCC(=O)NCC(=O)NCC(=O)NCC(=O)NCC(O)=O
InChI
1S/C10H17N5O6/c11-1-6(16)12-2-7(17)13-3-8(18)14-4-9(19)15-5-10(20)21/h1-5,11H2,(H,12,16)(H,13,17)(H,14,18)(H,15,19)(H,20,21)
InChI key
MXHCPCSDRGLRER-UHFFFAOYSA-N
Biochem/physiol Actions
Pentaglycine forms interpeptide bridges in peptidogylcan crosslinking and may be used as a tether for markers such as green fluorescent protein (GFP). Pentaglycine may be used as a model peptide in oligoglycine separation technology development.
Lagerklasse
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Christof Renner et al.
Molecular cancer, 9, 2-2 (2010-01-08)
It was previously demonstrated that the dipeptide carnosine inhibits growth of cultured cells isolated from patients with malignant glioma. In the present work we investigated whether carnosine also affects tumor growth in vivo and may therefore be considered for human
Hiroshi Tomoda
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 132(1), 37-44 (2012-01-05)
Methicillin-resistant Staphylococcus aureus (MRSA) is known as a major nosocomial pathogen that has also developed resistance to many antibiotics. Moreover, MRSA resistance to a last-resort antibiotic, vancomycin, has been reported. Therefore, new anti-infectious agents to prevent and treat MRSA infection
Aurelia Delaune et al.
PloS one, 6(2), e17054-e17054 (2011-03-10)
The WalKR two-component system is essential for viability of Staphylococcus aureus, a major pathogen. We have shown that WalKR acts as the master controller of peptidoglycan metabolism, yet none of the identified regulon genes explain its requirement for cell viability.
Global Trade Item Number
SKUGTIN
G5755-25MG04061832777559
G5755-1G04061826682715