Skip to Content
MilliporeSigma
F4638
Polaroid® black-and-white print film
Polaroid Type 667, This product has been discontinued. See product Z736627, Fujifilm FP-3000B, Instant Black & White Film
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41100000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
description
Format P, ISO Daylight Speed (ASA/DIN) 3000/36°, print size 31/4 in. 41/4 in.
packaging
pack of 10 exposures
manufacturer/tradename
Polaroid Type 667
General description
Medium contrast; high speed; coaterless; professional equivalent of Type107C.
These instant films are Print Only (P) or Print-plus-Negative (P/N). The negatives require brief clearing in sodium sulfite solutions. Unless otherwise noted, prints need to be coated. Films have panchromatic spectral sensitivity. Professional equivalents have the same characteristics as their counterparts, but meet more stringent photographic specifications.
Legal Information
Polaroid is a registered trademark of Polaroid Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library