Skip to Content
MilliporeSigma
Skip To
B9900
LC column replacement bed supports
column I.D. 1.5 cm
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
10 ea
Estimated to ship TODAYfromMILWAUKEE
$62.70
About This Item
NACRES:
SB.52
UNSPSC Code:
41115716
$62.70
Estimated to ship TODAYDetails
Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
technique(s)
LPLC: suitable
column I.D.
1.5 cm
General description
For use with flow adapter only.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
F9017SPK94100006R4778
technique(s)
LPLC: suitable
technique(s)
-
technique(s)
-
technique(s)
-
column I.D.
1.5 cm
column I.D.
1.0 cm
column I.D.
-
column I.D.
-
Still not finding the right product?
Explore all of our products under LC column replacement bed supports
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library