Skip to Content
MilliporeSigma
47614
Fluram
BioReagent, suitable for fluorescence, ≥99.0% (UV)
Synonym(s):
Fluorescamine, 4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H10O4
CAS Number:
Molecular Weight:
278.26
EC Number:
253-814-5
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
921143
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
product line
BioReagent
assay
≥99.0% (UV)
mp
153-157 °C
solubility
acetonitrile: soluble, ethanol: soluble
fluorescence
λex 234 nm, λex 390 nm; λem 480 nm in 0.5 M borate pH 8.5 (after derivatization with L-leucine)
suitability
suitable for fluorescence
SMILES string
O=C1OC2(OC=C(C2=O)c3ccccc3)c4ccccc14
InChI
1S/C17H10O4/c18-15-13(11-6-2-1-3-7-11)10-20-17(15)14-9-5-4-8-12(14)16(19)21-17/h1-10H
InChI key
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
Application
Fluram (fluorescamine) is a non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides and other molecules to form stable, highly fluorescent compounds. Fluorescamine is useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes and as a pre-column derivatization reagent. It effectively blocks newly generated amino termini in protein sequence analyses.
Non-fluorescent reagent that reacts readily under mild conditions with primary amines in amino acids and peptides to form stable, highly fluorescent compounds. Low background due to hydrolysis. Useful for the fluorometric assay of amino acids, protein, and proteolytic enzymes. Effectively blocks newly generated amino termini in protein sequence analyses.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Legal Information
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library