Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
PTFE plunger tip, cone tip, side port stainless steel needle (point style 5)
needle size
22 ga (side-port)
description
Gastight®
feature
needle type N (cemented)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81243
needle L
51 mm (2 in.)
volume
0.5 mL
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material PTFE plunger tip, cone tip, side port stainless steel needle (point style 5) | material PTFE plunger tip, cone tip, side port stainless steel needle (point style 5) | material PTFE plunger tip, cone tip, side port stainless steel needle (point style 5) | material PTFE plunger tip, cone tip, side port stainless steel needle (point style 5) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 81243 | manufacturer/tradename Hamilton 80039 | manufacturer/tradename Hamilton 80239 | manufacturer/tradename Hamilton 81139 |
| description Gastight® | description Gastight® | description Gastight® | description Gastight® |
| needle size 22 ga (side-port) | needle size 26s ga (side-port) | needle size 22s ga (side-port) | needle size 22s ga (side-port) |
| volume 0.5 mL | volume 10 μL | volume 25 μL | volume 250 μL |
Still not finding the right product?
Explore all of our products under Hamilton® GASTIGHT® Spritze, Serie 1700
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?