Skip to Content
MilliporeSigma
Skip To
PUMPKIT01
Pump Connection Kit
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 kit
About This Item
NACRES:
SB.51
UNSPSC Code:
41104214
This item is not able to be purchased as a guest user. For further information, please contact us at 800-244-1173.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 unit
General description
This water purification accessory is used to connect a pump to the high-flow outlet fitting of 30, 60 and 100 L polyethylene (PE) storage tanks.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
EZSWIRE01EZSKTMEM1ZSTWINLX2
packaging
pkg of 1 unit
packaging
pkg of 1 unit
packaging
-
packaging
pkg of 1 unit
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library