Skip to Content
MilliporeSigma
70634
pETBlue-1 Perfectly Blunt Cloning Kit - Novagen
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.51
UNSPSC Code:
41106617
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Quality Segment
Storage Class
10 - Combustible liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library