Skip to Content
MilliporeSigma
Skip To
SYNF321005
Synthware pear shaped flask
5 mL, joint: ST/NS 14/10
Synonym(s):
Synthware F321005, flask, pear shaped flask
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Available to ship TODAYfromMILWAUKEE
$24.30
About This Item
NACRES:
NB.51
UNSPSC Code:
41121804
Material:
borosilicate glass flask, pear-shaped , single-neck
Joint:
joint: ST/NS14/10
Packaging:
pkg of 1 ea
$24.30
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
borosilicate glass flask, pear-shaped , single-neck
joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F321005
capacity
5 mL , 50 mL , 5 mL
General description
Supplied with one compression cap, one Viton® O-ring and one PTFE –faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNF321010SYNF321025SYNF329250L
manufacturer/tradename
Synthware F321005
manufacturer/tradename
Synthware F321010
manufacturer/tradename
Synthware F321025
manufacturer/tradename
Synthware F329250L
material
borosilicate glass flask, single-neck , pear-shaped
material
borosilicate glass flask, pear-shaped , single-neck
material
borosilicate glass flask, pear-shaped , single-neck
material
borosilicate glass flask, pear-shaped , single-neck
capacity
5 mL , 5 mL, 50 mL
capacity
10 mL , 10 mL
capacity
25 mL , 25 mL
capacity
50 mL , 50 mL
joint
joint: ST/NS14/10
joint
joint: ST/NS14/10
joint
joint: ST/NS14/10
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Still not finding the right product?
Explore all of our products under Synthware pear shaped flask
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library