Merck
SYND192440
Synthware distilling receiver with hose connection
joint: ST/NS 24/40
Synonym(e):
Synthware D192440, glass distilling receiver
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D192440
Allgemeine Beschreibung
Cow-type design having three receivers spaced 40o apart. 105o angle between top outer joint and lower inner joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.