Merck
SYNA901420
Synthware Claisen distillation adapter with compression cap thermometer port
joint: ST/NS 14/20
Synonym(e):
Synthware 901420, Glass Distillation Adapter Claisen Thermometer
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A901420
Allgemeine Beschreibung
With threaded thermometer port, supplied complete with Nylon cap and Viton® O-ring to accommodate 6-7.5 mm O.D. thermometer.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company