Merck
M9602
Sigma-Aldrich
2-Methoxyacetophenon
95%
About This Item
Lineare Formel:
C6H5COCH2OCH3
CAS-Nummer:
Molekulargewicht:
150.17
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352100
PubChem Substanz-ID:
NACRES:
NA.22
Assay
95%
Form
liquid
Brechungsindex
n20/D 1.5365 (lit.)
bp
122-123 °C/18 mmHg (lit.)
mp (Schmelzpunkt)
7-8 °C (lit.)
Dichte
1.09 g/mL at 25 °C (lit.)
SMILES String
COCC(=O)c1ccccc1
InChI
1S/C9H10O2/c1-11-7-9(10)8-5-3-2-4-6-8/h2-6H,7H2,1H3
InChIKey
YRNDGUSDBCARGC-UHFFFAOYSA-N
Lagerklassenschlüssel
10 - Combustible liquids
WGK
WGK 3
Flammpunkt (°F)
215.6 °F - closed cup
Flammpunkt (°C)
102.00 °C - closed cup
Persönliche Schutzausrüstung
Eyeshields, Gloves
Die Dokumentenbibliothek aufrufen
Q Yin et al.
Journal of thrombosis and haemostasis : JTH, 14(3), 596-605 (2015-12-30)
ESSENTIALS: It is unclear whether interleukin-10 (IL-10) could affect clopidogrel metabolism and response. The bioactivation of and response to clopidogrel were determined between mice with or without IL-10. Maximum clopidogrel active metabolite levels were the major driver of platelet response
Henrique J Traesel et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 210, 82-97 (2018-11-18)
The conformational analysis of some 2‑(methoxy)‑2‑[(4‑substituted)‑phenylsulfanyl]‑(4'‑substituted) acetophenones was performed through infrared (IR) spectroscopic analysis of the carbonyl stretching band (νCO), supported by B3LYP/6-31+G(d,p) calculations and X-ray diffraction. Five (1-5) of the seven studied compounds (1-7) presented Fermi resonance (FR) on
Patrick Hemberger et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(19), 2217-2222 (2020-07-07)
Unveiling reaction mechanisms by isomer-selective detection of reactive intermediates requires advanced spectroscopic knowledge. We study the photoionization of fulvenone (c-C5 H4 =C=O), a reactive ketene species relevant in catalytic pyrolysis of lignin, which was generated by pyrolysis of 2-methoxy acetophenone.
Kobra Azizi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(25), 6439-6446 (2019-03-19)
Manganese(III) porphyrin chloride complexes have been developed for the first time as catalysts for the acceptorless dehydrogenative coupling of alcohols and amines. The reaction has been applied to the direct synthesis of imines, tertiary amines and quinolines where only hydrogen
Marcela L Contreras et al.
Frontiers in neuroscience, 11, 338-338 (2017-07-01)
Ethanol exposure increases oxidative stress in developing organs, including the brain. Antioxidant treatment during maternal ethanol ingestion improves behavioral deficits in rodent models of fetal alcohol spectrum disorder (FASD). However, the impact of general antioxidant treatment in their adult offspring