CLS6383500
Pyrex® cylindrical separatory funnel, PTFE stopcock
Synonym(e):
Corning® 6383
Empfohlene Produkte
Verbindung
joint stopper27
Hersteller/Markenname
Corning 6383-500
Kapazität
500 mL
Absperrhahngröße
2
Verwandte Kategorien
Allgemeine Beschreibung
Graduated, with ST Pyrex stopper and microfinish PTFE stopcock.
CLS6383A100 - does not include a stopper
CLS6383A100 - does not include a stopper
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.