Merck
CLS5400500
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 500 mL
Synonym(e):
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
wide-mouth
Verbindung
joint ST/NS stopper22
Sterilität
non-sterile
Leistungsmerkmale
cap: no
closure type ST stopper
Verpackung
case of 6
Hersteller/Markenname
Corning 5400-500
Kapazität
500 mL
Allgemeine Beschreibung
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.