MilliporeSigma
LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel Que Z-Sep/C18 QuEChERS sorbent
LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel™ Que Z-Sep/C18 QuEChERS sorbent application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
SPE tube or plate
Product No.
Description
Pricing
Supel QuE
Citrate Extraction Tube, pk of 50, suitable for EN 15662
Supel QuE
Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
related product
Product No.
Description
Pricing
Supel QuE
Empty Centrifuge Tube with Lid, centrifuge tube volume 50 mL, pk of 50, suitable for EN 15662:2008 per BS
Supel QuE
PSA Tube, pack of 100, AOAC® 2007.01
Supel QuE
PSA/C18 Tube, pack of 100, AOAC® 2007.01
standard
Product No.
Description
Pricing
Mecarbam
PESTANAL®, analytical standard
2-Octyl-4-isothiazolin-3-one
PESTANAL®, analytical standard
Aldicarb
PESTANAL®, analytical standard
Etrimfos
PESTANAL®, analytical standard
Fenoxycarb
PESTANAL®, analytical standard
Trichlorfon
PESTANAL®, analytical standard
Iprobenfos
PESTANAL®, analytical standard
Terbufos
PESTANAL®, analytical standard
Phosmet
PESTANAL®, analytical standard
EPN
PESTANAL®, analytical standard
Dibrom®
PESTANAL®, analytical standard
Parathion-methyl
PESTANAL®, analytical standard
Pyraclostrobin
PESTANAL®, analytical standard
Spiromesifen
PESTANAL®, analytical standard
Flufenoxuron
PESTANAL®, analytical standard
Fenpyroximate
PESTANAL®, analytical standard
Lufenuron
PESTANAL®, analytical standard
Fenhexamid
PESTANAL®, analytical standard
Tolclofos-methyl
PESTANAL®, analytical standard
CONDITIONS
sample preparation
Dispersive (QuEChERS)
sample/matrix
10 g of pureed oranges (homogenized with rind); spike at 50 ppb (add 16.75 μL of a custom made pesticide mix, each analyte at 30 μg/mL)
extraction tube
Supel QuE Citrate (EN) Tube (55227-U)
extraction process
add 10 mL acetonitrile; shake for 1 minute; add contents of a Supel QuE citrate extraction tube (55227-U); shake immediately for 1 minute; centrifuge at 3200 rpm for 5 minutes
clean-up tube
Supel-Q PSA/C18 (55284-U)
clean-up process
transfer 0.7 mL of the acetonitrile layer into a Supel QuE Z-Sep/C18 cleanup tube (55284-U); shake for 1 minute; centrifuge at 5000 rpm for 5 minutes; (transfer 0.2 mL of the supernatant into an empty 1.5 mL centrifuge tube; add 0.2 mL of water; centrifuge at 5000 rpm for 2 minutes.)
column
Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 μm particles (53822-U)
mobile phase
[A] 10 mM ammonium acetate in water; [B] 10 mM ammonium acetate in acetonitrile
gradient
Hold at 30% B for 1 min; 30% to 80% B in 2 min; hold at 80% B for 4 min; hold at 100% B for 3 min; hold at 30% B for 3 min
flow rate
0.3 mL/min
pressure
2730 psi (188 bar)
column temp.
30 °C
detector
MS/MS, ESI positive
Description
Analysis Note
This demonstrates the application of the Z-Sep/C18 sorbent to the cleanup of fruits in the LC-MS-MS analysis of polar pesticides including organophosphates, carbamates and benzoylureas
Sample pretreatment: 10 g of pureed oranges homogenized with rind; spike at 50 ppb by adding 16.75 μL of a pesticide mix containing each analyte at 30 μg/mL
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC
null