MilliporeSigma
HPLC Analysis of Proteins on Proteomix® SCX Columns, Effect of Particle Size of Resolution
HPLC Analysis of Proteins on Proteomix® SCX Columns, Effect of Particle Size of Resolution application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SCX-NP10 Strong Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 10 μm particle size, stainless steel
Proteomix® SCX-NP1.7 Strong Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 1.7 μm particle size, stainless steel
Proteomix® SCX-NP3 Strong Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 3 μm particle size, stainless steel
Proteomix® SCX-NP5 Strong Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 5 μm particle size, stainless steel
standard
Product No.
Description
Pricing
Lysozyme from chicken egg white
powder or granules, ≥90 %, ≥39,000 units/mg protein
Cytochrome c from equine heart
≥95% (SDS-PAGE)
Ribonuclease A from bovine pancreas
for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
CONDITIONS
column
Proteomix SCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 μm particles (Z777146)
column
Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777151)
mobile phase
(A) 20 mM sodium phosphate buffer, pH 6.0, (B) A + 1.0 M NaCl
gradient
0-25 min from 0-75% B, 15 min prewash
flow rate
0.75 mL/min
column temp.
Ambient
detector
214 nm
injection
5 μL
sample
each 1 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies