MilliporeSigma
HPLC Analysis of Polyphenols in Nero d′Avola Red Wine on Discovery® HS C18 (UV 354 nm)
HPLC Analysis of Polyphenols in Nero d’Avola Red Wine on Discovery® HS C18 (UV 354 nm) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
3 μm particle size, L × I.D. 5 cm × 2.1 mm
standard
Product No.
Description
Pricing
Rutin hydrate
≥94% (HPLC), powder
Isoquercitrin
primary reference standard
Caftaric acid
analytical standard
CONDITIONS
sample/matrix
Nero d’Avola wine sample filtered prior injection through a 0.45 μm nylon membrane
column
Discovery HS C18, 15 cm x 2.1 mm I.D., 3 μm particles (569255-U)
mobile phase
[A] water, pH 3 with fomic acid; [B] Acetonitrile, pH 3 with fomic acid
flow rate
0.2 mL/min
column temp.
30 °C
detector
UV, 354 nm
injection
5 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany