MilliporeSigma
HPLC Analysis of Polyphenols in Nero d′Avola Red Wine on Discovery® HS C18 (UV 280 nm)
HPLC Analysis of Polyphenols in Nero d’Avola Red Wine on Discovery® HS C18 (UV 280 nm) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
3 μm particle size, L × I.D. 5 cm × 2.1 mm
standard
Product No.
Description
Pricing
Caffeic acid
≥98.0% (HPLC)
Rutin hydrate
≥94% (HPLC), powder
Ethyl gallate
antioxidant, ≥96.0% (HPLC)
Procyanidin B2
analytical standard
Epicatechin solution
certified reference material, 2000 μg/mL in methanol, ampule of 0.5 mL
Isoquercitrin
primary reference standard
(+)-Catechin
analytical standard
Gallic acid
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Caftaric acid
analytical standard
CONDITIONS
sample/matrix
Nero d’Avola wine sample filtered prior injection through a 0.45 μm nylon membrane
column
Discovery HS C18, 15 cm x 2.1 mm I.D., 3 μm particles (569255-U)
mobile phase
[A] water, pH 3 with formic acid; [B] acetonitrile, pH 3 with formic acid
flow rate
0.2 mL/min
column temp.
30 °C
detector
UV, 280 nm
injection
5 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany