MilliporeSigma
HPLC Analysis of Phenylephrine Enantiomers on Astec® CYCLOBOND I 2000 AC
HPLC Analysis of Phenylephrine Enantiomers on Astec® CYCLOBOND I 2000 AC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 AC Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
(R)-(−)-Phenylephrine hydrochloride
analytical standard
(R)-(−)-Phenylephrine hydrochloride
powder
(R)-(−)-Phenylephrine hydrochloride
tested according to Ph. Eur.
CONDITIONS
column
CYCLOBOND I 2000 AC, 25 cm x 4.6 mm I.D., 5 μm particles (20124AST)
mobile phase
[A] 0.5% w/v sodium acetate, pH 5.5; [B] methanol; (90:10, A:B)
flow rate
0.5 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null