MilliporeSigma
HPLC Analysis of Oligonucleotides on Proteomix® SAX
HPLC Analysis of Oligonucleotides on Proteomix® SAX application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SAX-NP5 Strong Anion Exchange Column
L × I.D. 25 cm × 4.6 mm, 5 μm particle size
CONDITIONS
column
Proteomix SAX-NP5, 25 cm x 4.6 mm I.D., 5 μm particles (Z777229)
mobile phase
A: 20 mM Tris, pH 8.0 B: A + 0.5 M NaCl
gradient
0 - 100% B in 30 minutes
flow rate
0.5 mL/min
pressure
1073 psi (74 bar)
column temp.
25 °C
detector
UV 260 nm
injection
5 μL
sample
(10 mM each oligonucleotide in water)
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies