MilliporeSigma
HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
standard
Product No.
Description
Pricing
D-Methionine
≥98% (HPLC)
L-Methionine
from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
L-Methionine
reagent grade, ≥98% (HPLC)
DL-Methionine
BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
L-Methionine
L-Methionine
BioUltra, ≥99.5% (NT)
DL-Methionine
≥99.0% (NT)
DL-Methionine
99%, FCC, FG
L-Methionine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
(A) water; (B) methanol; (C) formic acid; (30:70:0.02, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
300 μg /mL in 30:70 water:methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC