MilliporeSigma
HPLC Analysis of MAb on Proteomix® SCX-NP5 Comparing Different Buffer Systems and pH Gradients
HPLC Analysis of MAb on Proteomix® SCX-NP5 Comparing Different Buffer Systems and pH Gradients application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SCX-NP5 Strong Cation Exchange Column
L × I.D. 25 cm × 4.6 mm, 5 μm particle size, stainless steel
CONDITIONS
column
Proteomix SCX-NP5, 25 cm x 4.6 mm I.D., 5 μm particles (Z777161)
mobile phase
Mobile phase A: (A) 20 mM MES, pH 6.0; (B) A + 1 M NaCl, pH 6.0Mobile phase B: (A) 2.4 mM Tris, 1.5 mM imidazole, 11.6 mM piperazine, pH 6.0; (B) A, pH 10.5
gradient
Gradient A: 7% B held for 2 min; to 14% B in 20 min, Gradient B: 0 to 30% B in 2 min; to 55% B in 30 min
flow rate
0.8 mL/min
detector
UV, 280 nm
injection
5 μL
sample
10 mg/mL
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies