HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
![HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/293/671/e6cfcc56-fe3a-401e-897b-1b3c6d98e5b8/800/e6cfcc56-fe3a-401e-897b-1b3c6d98e5b8.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] ammonium hydroxide; (99.88:0.1:0.01, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC