MilliporeSigma
HPLC Analysis of Acidic Herbicides in Water on a Polymeric C18 Column after SPE using Supelclean ENVI-Carb
HPLC Analysis of Acidic Herbicides in Water on a Polymeric C18 Column after SPE using Supelclean™ ENVI™-Carb application for SPE, application for HPLC
Materials
SPE tube or plate
Product No.
Description
Pricing
Supelclean ENVI-Carb SPE Tube
bed wt. 250 mg, volume 6 mL, pk of 30
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
Fresh 1 L water samples, dechlorinated with sodium thiosulfate when necessary, at ambient temperature and pH.
SPE tube/cartridge
Supelclean ENVI-Carb, 250 mg/6 mL (57092)
condition
10 mL DI water at 20 mL/min
sample addition
0.9 L water sample at 20 mL/min
drying
10 min using clean nitrogen
washing
10 mL DI water at 20 mL/min
elution
10 mL 0.1% phosphoric acid in methylene chloride:acetonitrile (80:20) at 5 mL/min
column
polymeric-coated silica-based PAH specialty column, 20 cm × 3 mm I.D., 5 μm (Supelco equivalent, SUPELCOSIL LC-PAH, available upon request)
mobile phase
gradient, [A]: 0.05% phosphoric acid in DI water; [B]: acetonitrile
flow rate
0.5 mL/min
column temp.
50 °C
detector
photodiode array- peak width: 0.053 min, sampling interval: 0.320 sec, monitor 210 nm & 225 nm
injection
10 μL of extract (4-5 ppb each analyte in water)
Description
Application
using Zymark AutoTrace Extraction WorkStation 1.20
Legal Information
ENVI is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC