MilliporeSigma
GC Analysis of (R)-(+)-1-Phenylethanol on Astec® CHIRALDEX B-PM
GC Analysis of (R)-(+)-1-Phenylethanol on Astec® CHIRALDEX™ B-PM suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PM Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(R)-(+)-1-Phenylethanol
for chiral derivatization, LiChropur, ≥99.0%
(R)-(+)-1-Phenylethanol
≥98.5% (sum of enantiomers, GC)
(R)-1-Phenylethanol
97%
CONDITIONS
column
Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 μm (76023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
1 μL, 80:1 split
sample
3 mg/mL in methylene chloride
peak 1: (R)-(+)-1-phenylethanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC