MilliporeSigma
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX G-DP
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DP suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-DP Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
Application
Product No.
Description
Pricing
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX G-PN
suitable for GC
standard
Product No.
Description
Pricing
D-Amphetamine hemisulfate salt solution
1.0 mg/mL in methanol, analytical standard, for drug analysis
DL-Amphetamine sulfate salt
D-Amphetamine hemisulfate salt
DL-Amphetamine hydrochloride
analytical standard
CONDITIONS
column
Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven
140 °C
detector
FID, 250 °C
inj. temp.
250 °C
carrier gas
helium, 2.7 mL/min., constant flow
injection
1 μL, 80:1 split
liner
4 mm I.D., FocusLiner
sample
4 mg/mL each compound in methylene chloride
peak 1: d-amphetamine (N-TFA derivative)
peak 2: l-amphetamine (N-TFA derivative)
Description
Analysis Note
Elution order reversal (l,d) by using CHIRALDEX G-PN, 130°C (see G005053).
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC