MilliporeSigma
  • Quantitative characterization of food products by two-dimensional D-T2 and T1-T2 distribution functions in a static gradient.
Quantitative characterization of food products by two-dimensional D-T2 and T1-T2 distribution functions in a static gradient.
Journal of colloid and interface science (2005-11-23)
Martin D Hürlimann, Lauren Burcaw, Yi-Qiao Song
ABSTRACT
We present new NMR techniques to characterize food products that are based on the measurement of two-dimensional diffusion-T2 relaxation and T1-T2 relaxation distribution functions. These measurements can be performed in magnets of modest strength and low homogeneity and do not require pulsed gradients. As an illustration, we present measurements on a range of dairy products that include milks, yogurt, cream, and cheeses. The two-dimensional distribution functions generally exhibit two distinct components that correspond to the aqueous phase and the liquid fat content. The aqueous phase exhibits a relatively sharp peak, characterized by a large T1/T2 ratio of around 4. The diffusion coefficient and relaxation times are reduced from the values for bulk water by an amount that is sample specific. The fat signal has a similar signature in all samples. It is characterized by a wide T2 distribution and a diffusion coefficient of 10(-11) m2/s for a diffusion time of 40 ms, determined by bounded diffusion in the fat globules of 3 microm diameter.
Product Description
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.10 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm