MilliporeSigma
Photochemical production of conjugated linoleic acid from soybean oil.
Lipids (2004-11-24)
R R Gangidi, A Proctor
ABSTRACT
Conjugated linoleic acid (CLA), an anticarcinogenic compound with numerous other health benefits, is present mainly in dairy and beef lipids. The main CLA isomer present in dairy and beef lipids is cis 9, trans 11 CLA at a 0.5% concentration. The typical minimum human dietary intake of CLA is 10 times less than the 3 g/d suggested requirement that has been extrapolated from animal and cell-line studies. The objectives of this study were to produce CLA isomers from soybean oil by photoisomerization of soybean oil linoleic acid and to study the oxidation status of the oil. Refined, bleached, and deodorized soybean oil with added iodine concentrations of 0, 0.1, 0.25, and 0.5% was exposed to a 100-W mercury lamp for 0 to 120 h. An SP-2560 fused-silica capillary GC column with FID was used to analyze the esterified CLA isomers in the photoisomerized oil. The CLA content of the individual isomers was optimized by response surface methodology. Attenuated total reflectance (ATR)-FTIR spectra in the 3400 to 3600 cm(-1) range and 1H NMR spectra in the 8 to 12 ppm range of the photoisomerized soybean oil were obtained to follow hydroperoxide formation. The largest amount of cis 9, trans 11 CLA isomer in soybean oil was 0.6%, obtained with 0.25% iodine and 84 h of photoisomerization. Lipid hydroperoxide peaks in the ATR-FTIR spectra and aldehyde peaks in the 1H NMR spectra were not observed in the photoisomerized soybean oil, and the spectra were similar to that of fresh soybean oil. This study shows that CLA isomers can be produced simply and inexpensively from soybean oil by photoisomerization.
Product Description
Supelco
SP®-2560 Capillary GC Column, L × I.D. 75 m × 0.18 mm, df 0.14 μm
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2560 Capillary GC Column, L × I.D. 200 m × 0.25 mm, df 0.20 μm