MilliporeSigma
  • Exploring boar sperm sialylation during capacitation using boronic acid-functionalized beads.
Exploring boar sperm sialylation during capacitation using boronic acid-functionalized beads.
Reproduction (Cambridge, England) (2017-12-23)
Yuanxian Wang, Yihua Gu, Huihui Gao, Yao Gao, Jianhang Shao, Weijun Pang, Wuzi Dong
ABSTRACT
Sialic acid (SA), which usually occupies the terminal position of oligosaccharide chains in mammalian spermatozoa, has important functions in fertilization. Compared with other methods, such as lectin probing, boronic acid could recognize and bind SA with a higher affinity and specificity at pH 6.9. In this study, two boronic acid carriers, 3-aminophenylboronic acid-labeled fluorescent latex (CML-APBA) and magnetic beads (CMM-APBA were applied to explore surface sialylation profile and sialoglycoproteins of the boar sperm. There are three binding sections of CML-APBA on the head of ejaculated sperm: acrosomal region, equatorial segment and the head posterior, which are the major regions undergoing sialylation. After capacitation
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green, aqueous suspension, 0.03 μm mean particle size
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)