MilliporeSigma
An Approach to Discovering Novel Muscarinic M
The Journal of pharmacology and experimental therapeutics (2017-10-14)
Emi Kurimoto, Satoru Matsuda, Yuji Shimizu, Yuu Sako, Takao Mandai, Takahiro Sugimoto, Hiroki Sakamoto, Haruhide Kimura
ABSTRACT
Activation of muscarinic M
Sigma-Aldrich
BQCA, ≥98% (HPLC)