MilliporeSigma
  • Acquired resistance to tyrosine kinase inhibitors may be linked with the decreased sensitivity to X-ray irradiation.
Acquired resistance to tyrosine kinase inhibitors may be linked with the decreased sensitivity to X-ray irradiation.
Oncotarget (2018-02-13)
Maxim Sorokin, Roman Kholodenko, Anna Grekhova, Maria Suntsova, Margarita Pustovalova, Natalia Vorobyeva, Irina Kholodenko, Galina Malakhova, Andrew Garazha, Artem Nedoluzhko, Raif Vasilov, Elena Poddubskaya, Olga Kovalchuk, Leila Adamyan, Vladimir Prassolov, Daria Allina, Denis Kuzmin, Kirill Ignatev, Andreyan Osipov, Anton Buzdin
ABSTRACT
Acquired resistance to chemotherapy and radiation therapy is one of the major obstacles decreasing efficiency of treatment of the oncologic diseases. In this study, on the two cell lines (ovarian carcinoma SKOV-3 and neuroblastoma NGP-127), we modeled acquired resistance to five target anticancer drugs. The cells were grown on gradually increasing concentrations of the clinically relevant tyrosine kinase inhibitors (TKIs) Sorafenib, Pazopanib and Sunitinib, and rapalogs Everolimus and Temsirolimus, for 20 weeks. After 20 weeks of culturing, the half-inhibitory concentrations (IC50) increased by 25 - 186% for the particular combinations of the drugs and cell types. We next subjected cells to 10 Gy irradiation, a dose frequently used in clinical radiation therapy. For the SKOV-3, but not NGP-127 cells, for the TKIs Sorafenib, Pazopanib and Sunitinib, we noticed statistically significant increase in capacity to repair radiation-induced DNA double strand breaks compared to naïve control cells not previously treated with TKIs. These peculiarities were linked with the increased activation of ATM DNA repair pathway in the TKI-treated SKOV-3, but not NGP-127 cells. Our results provide a new cell culture model for studying anti-cancer therapy efficiency and evidence that there may be a tissue-specific radioresistance emerging as a side effect of treatment with TKIs.
Sigma-Aldrich
Temsirolimus, ≥98% (HPLC)
Sigma-Aldrich
Complete Whole Transcriptome Amplification Kit, DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias
Sigma-Aldrich
Anti-ATM phosphoSer1981 Antibody, clone 10H11.E12, clone 10H11.E12, Chemicon®, from mouse