MilliporeSigma
  • The Transcription Factor STAT6 Mediates Direct Repression of Inflammatory Enhancers and Limits Activation of Alternatively Polarized Macrophages.
The Transcription Factor STAT6 Mediates Direct Repression of Inflammatory Enhancers and Limits Activation of Alternatively Polarized Macrophages.
Immunity (2018-01-19)
Zsolt Czimmerer, Bence Daniel, Attila Horvath, Dominik Rückerl, Gergely Nagy, Mate Kiss, Matthew Peloquin, Marietta M Budai, Ixchelt Cuaranta-Monroy, Zoltan Simandi, Laszlo Steiner, Bela Nagy, Szilard Poliska, Csaba Banko, Zsolt Bacso, Ira G Schulman, Sascha Sauer, Jean-Francois Deleuze, Judith E Allen, Szilvia Benko, Laszlo Nagy
ABSTRACT
The molecular basis of signal-dependent transcriptional activation has been extensively studied in macrophage polarization, but our understanding remains limited regarding the molecular determinants of repression. Here we show that IL-4-activated STAT6 transcription factor is required for the direct transcriptional repression of a large number of genes during in vitro and in vivo alternative macrophage polarization. Repression results in decreased lineage-determining transcription factor, p300, and RNA polymerase II binding followed by reduced enhancer RNA expression, H3K27 acetylation, and chromatin accessibility. The repressor function of STAT6 is HDAC3 dependent on a subset of IL-4-repressed genes. In addition, STAT6-repressed enhancers show extensive overlap with the NF-κB p65 cistrome and exhibit decreased responsiveness to lipopolysaccharide after IL-4 stimulus on a subset of genes. As a consequence, macrophages exhibit diminished inflammasome activation, decreased IL-1β production, and pyroptosis. Thus, the IL-4-STAT6 signaling pathway establishes an alternative polarization-specific epigenenomic signature resulting in dampened macrophage responsiveness to inflammatory stimuli.
Product Description
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20