MilliporeSigma
Pterostilbene 4'-
Oxidative medicine and cellular longevity (2017-06-14)
Yingqing Chen, Jeongmin Park, Yeonsoo Joe, Hyeok-Jun Park, Seung-Joo Jekal, Daisuke Sato, Hiroki Hamada, Hun Taeg Chung
ABSTRACT
Pterostilbene, a dimethyl ester analog of resveratrol, has anti-inflammatory and antioxidative effects and alters cell proliferation. Tristetraprolin (TTP) promotes the degradation of proinflammatory mediators via binding to adenosine and uridine- (AU-) rich elements (ARE) located in the 3'-untranslated regions of mRNAs. Here, we utilized pterostilbene 4'-
Sigma-Aldrich
Anti-TTP (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution