MilliporeSigma
  • 3D Bioprinting of Low-Concentration Cell-Laden Gelatin Methacrylate (GelMA) Bioinks with a Two-Step Cross-linking Strategy.
3D Bioprinting of Low-Concentration Cell-Laden Gelatin Methacrylate (GelMA) Bioinks with a Two-Step Cross-linking Strategy.
ACS applied materials & interfaces (2018-02-07)
Jun Yin, Mengling Yan, Yancheng Wang, Jianzhong Fu, Hairui Suo
ABSTRACT
Methacrylated gelatin (GelMA) has been widely used as a tissue-engineered scaffold material, but only low-concentration GelMA hydrogels were found to be promising cell-laden bioinks with excellent cell viability. In this work, we reported a strategy for precise deposition of 5% (w/v) cell-laden GelMA bioinks into controlled microarchitectures with high cell viability using extrusion-based three-dimensional (3D) bioprinting. By adding gelatin into GelMA bioinks, a two-step cross-linking combining the rapid and reversible thermo-cross-linking of gelatin with irreversible photo-cross-linking of GelMA was achieved. The GelMA/gelatin bioinks showed significant advantages in processability because the tunable rheology and the rapid thermo-cross-linking of bioinks improved the shape fidelity after bioprinting. Here, the rheology, mechanical properties, and swelling of GelMA/gelatin bioinks with different concentration ratios were carefully characterized to obtain the optimized bioprinting setup. We successfully printed the 5% (w/v) GelMA with 8% (w/v) gelatin into 3D structures, which had the similar geometrical resolution as that of the structures printed by 30% (w/v) GelMA bioinks. Moreover, the cell viability of 5/8% (w/v) GelMA/gelatin bioinks was demonstrated by in vitro culture and cell printing of bone marrow stem cells (BMSCs). Larger BMSC spreading area was found on 5/8% (w/v) GelMA/gelatin scaffolds, and the BMSC viability after the printing of 5/8% (w/v) GelMA/gelatin cell-laden bioinks was more than 90%, which was very close to the viability of printing pure 5% (w/v) GelMA cell-laden bioinks. Therefore, this printing strategy of GelMA/gelatin bioinks may extensively extend the applications of GelMA hydrogels for tissue engineering, organ printing, or drug delivery.
Sigma-Aldrich
TissueFab® - low endotoxin GelMA-UV bioink, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -UV/365 nm