MilliporeSigma
  • Hydrophobic boron compound-loaded poly(l-lactide-co-glycolide) nanoparticles for boron neutron capture therapy.
Hydrophobic boron compound-loaded poly(l-lactide-co-glycolide) nanoparticles for boron neutron capture therapy.
Colloids and surfaces. B, Biointerfaces (2017-08-15)
Issei Takeuchi, Kensuke Nomura, Kimiko Makino
ABSTRACT
Poly(DL-lactide-co-glycolide) (PLGA) has been widely used and studied because of its biocompatibility and biodegradability. Recently, the usefulness of nanoparticles using poly(L-lactide-co-glycolide) (PLLGA) having a higher glass transition temperature than PLGA was suggested. In this study, we investigated the availability of boron compound-loaded PLGA and PLLGA nanoparticles for boron neutron capture therapy (BNCT) by conducting biodistribution study using tumor-bearing mice. o-Carborane, a hydrophobic boron compound, was used as a boron carrier, and o-carborane-albumin conjugate was used as a control. We prepared PLGA and PLLGA nanoparticles with diameters of 100nm and 150nm. In 100-nm PLLGA nanoparticles, the boron concentration in the tumor reached 113.9±15.8μg/g of tissue at 8h after administration. This result indicated that 100-nm PLLGA nanoparticles were able to achieve an intratumoral
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
o-Carborane, 98%