MilliporeSigma
Involvement of H
Pesticide biochemistry and physiology (2017-12-01)
Xiaoyong Luo, Zhihang Liu, Yukari Sunohara, Hiroshi Matsumoto, Pingliang Li
ABSTRACT
In order to understand the action mechanism of fluazifop-P-butyl (FB) in bristly starbur (Acanthospermum hispidum D.C.), a susceptible plant, the role of active oxygen species (ROS) in herbicide-induced cell death in shoots was investigated. FB-induced phytotoxicity was not reduced by the antioxidants, 1,4-diazabicyclooctane (dabaco), sodium azide, l-tryptophan, d-tryptophan, hydroquinone and dimethyl pyridine N-oxide (DMPO). The activities of superoxide dismutase (SOD) and catalase (CAT), in bristly starbur seedlings were significantly increased by FB at 12 HAT and 24 HAT, while ascorbate peroxidase (APX) and glutathione reductase (GR) activities increased only at 12 HAT. The contents of H
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Supelco
Fluazifop-P-butyl, PESTANAL®, analytical standard
Sigma-Aldrich
2,3-Lutidine, 99%