MilliporeSigma
  • DFT computations on: Crystal structure, vibrational studies and optical investigations of a luminescent self-assembled material.
DFT computations on: Crystal structure, vibrational studies and optical investigations of a luminescent self-assembled material.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-10-19)
A Kessentini, A Ben Ahmed, T Dammak, M Belhouchet
ABSTRACT
The current work undertakes the growth and the physicochemical properties of a novel green-yellow luminescence semi-organic material, the 3-picolylammonium bromide abbreviated (Pico-Br). In this paper, we report the X-ray diffraction measurements which show that the crystal lattice consists of distinct 3-picolylammonium cations and free bromide anions connected via NH⋯Br and NH⋯N hydrogen bonds leading to form a two dimensional frameworks. Molecular geometry compared with its optimized counterpart shows that the quantum chemical calculations carried out with density functional method (DFT) well produce the perceived structure by X-ray resolution of the studied material. To provide further insight into the spectroscopic properties, additional characterization of this material have been performed with Raman and infrared studies at room temperature. Theoretical computations have been computed using the (DFT) method at B3LYP/LanL2DZ level of theory implemented within Gaussian 03 program to study the vibrational spectra of the investigated molecule in the ground state. Optical absorption spectrum inspected by UV-visible absorption reveals the appearance of sharp optical gap of 280nm (4.42eV) as well as a strong green photoluminescence emission at 550nm (2.25eV) is detected on the photoluminescence (PL) spectrum at room temperature. Using the TD/DFT method, HOMO-LUMO energy gap and the Mulliken atomic charges were calculated in order to get an insight into the material. Good agreement between the theoretical results and the experimental ones was predicted.
Sigma-Aldrich
3-Picolylamine, ≥99%