MilliporeSigma
  • N-phenylpropyl-N'-substituted piperazines occupy sigma receptors and alter methamphetamine-induced hyperactivity in mice.
N-phenylpropyl-N'-substituted piperazines occupy sigma receptors and alter methamphetamine-induced hyperactivity in mice.
Pharmacology, biochemistry, and behavior (2016-11-17)
Dennis K Miller, Eric S Park, Susan Z Lever, John R Lever
ABSTRACT
This study examined the effect of the N-phenylpropyl-N'-substituted piperazine ligands SA4503 (3.4-dimethoxyphenethyl), YZ-067 (4-methoxyphenethyl), YZ-185 (3-methoxyphenethyl) and Nahas-3h (4-methoxybenzyl) on methamphetamine-induced hyperactivity in mice. In a previous study in rats, SA4503 increased methamphetamine-induced hyperactivity at a lower ligand dose and enhanced it at a higher dose. The other ligands have not been investigated in this assay. Presently, mice were administered sigma ligands, and specific [
Sigma-Aldrich
SA4503, ≥98% (HPLC)