MilliporeSigma
Anti-aggregant tau mutant promotes neurogenesis.
Molecular neurodegeneration (2017-12-06)
Maria Joseph, Marta Anglada-Huguet, Katharina Paesler, Eckhard Mandelkow, Eva-Maria Mandelkow
ABSTRACT
The microtubule-associated protein Tau plays a role in neurodegeneration as well as neurogenesis. Previous work has shown that the expression of the pro-aggregant mutant Tau repeat domain causes strong aggregation and pronounced neuronal loss in the hippocampus whereas the anti-aggregant form has no deleterious effects. These two proteins differ mainly in their propensity to form ß structure and hence to aggregate. To elucidate the basis of these contrasting effects, we analyzed organotypic hippocampal slice cultures (OHSCs) from transgenic mice expressing the repeat domain (RD) of Tau with the anti-aggregant mutation (Tau The expression of the anti-aggregant Tau The data suggest that the expression of anti-aggregant Tau
Sigma-Aldrich
AIDA, solid