MilliporeSigma
  • Altered metabolic landscape in IDH-mutant gliomas affects phospholipid, energy, and oxidative stress pathways.
Altered metabolic landscape in IDH-mutant gliomas affects phospholipid, energy, and oxidative stress pathways.
EMBO molecular medicine (2017-10-22)
Fred Fack, Saverio Tardito, Guillaume Hochart, Anais Oudin, Liang Zheng, Sabrina Fritah, Anna Golebiewska, Petr V Nazarov, Amandine Bernard, Ann-Christin Hau, Olivier Keunen, William Leenders, Morten Lund-Johansen, Jonathan Stauber, Eyal Gottlieb, Rolf Bjerkvig, Simone P Niclou
ABSTRACT
Heterozygous mutations in NADP-dependent isocitrate dehydrogenases (IDH) define the large majority of diffuse gliomas and are associated with hypermethylation of DNA and chromatin. The metabolic dysregulations imposed by these mutations, whether dependent or not on the oncometabolite D-2-hydroxyglutarate (D2HG), are less well understood. Here, we applied mass spectrometry imaging on intracranial patient-derived xenografts of IDH-mutant versus IDH wild-type glioma to profile the distribution of metabolites at high anatomical resolution
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
9-Aminoacridine, ≥99.5% (HPLC)