MilliporeSigma
  • Cell micropatterning reveals the modulatory effect of cell shape on proliferation through intracellular calcium transients.
Cell micropatterning reveals the modulatory effect of cell shape on proliferation through intracellular calcium transients.
Biochimica et biophysica acta (2017-10-01)
Jie Tong, Ying Qi, Xiangmiao Wang, Liyin Yu, Chang Su, Wenjun Xie, Jianbao Zhang
ABSTRACT
The mechanism by which cell shape regulates the function of the cell is one of the most important biological issues, but it remains unclear. Here, we investigated the effect of the regulation of cell shape on proliferation by using a micropatterning approach to confine MC3T3-E1 cells into specific shapes. Our results show that the proliferation rate for rectangle-, triangle-, square- and circle-shaped osteoblasts increased sequentially and was related to the nuclear shape index (NSI) but not the cell shape index (CSI). Interestingly, intracellular calcium transients also displayed different patterns, with the number of Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP2A2
Sigma-Aldrich
MISSION® esiRNA, targeting human ITPR1