MilliporeSigma
  • Autonomous and Continuous Stimuli-Responsive Polymer Surface for Antibacterial Application through Enzymatic Self-Propagating Reactions.
Autonomous and Continuous Stimuli-Responsive Polymer Surface for Antibacterial Application through Enzymatic Self-Propagating Reactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-05)
Chao Ding, Zhengqing Yan, Jinsong Ren, Xiaogang Qu
ABSTRACT
Stimuli-responsive polymer materials inspired by biological materials have invoked increasing research interest; however, they still suffer from limitations such as finite amplified responses and poor sensitivity of the unstimulated parts. Herein, a new strategy for creating H
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
Sigma-Aldrich
3-Aminophenylboronic acid hydrochloride, 98%