MilliporeSigma
Bcl-x
Mucosal immunology (2017-04-13)
X Zhao, N Khan, H Gan, F Tzelepis, T Nishimura, S-Y Park, M Divangahi, H G Remold
ABSTRACT
Virulent Mycobacterium tuberculosis (Mtb) triggers necrosis in host Mϕ, which is essential for successful pathogenesis in tuberculosis. Here we demonstrate that necrosis of Mtb-infected Mϕ is dependent on the action of the cytosolic Receptor Interacting Protein Kinase 3 (RIPK3) and the mitochondrial Bcl-2 family member protein B-cell lymphoma-extra large (Bcl-x
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution