MilliporeSigma
  • CETP: Pharmacogenomics-Based Response to the CETP Inhibitor Dalcetrapib.
CETP: Pharmacogenomics-Based Response to the CETP Inhibitor Dalcetrapib.
Arteriosclerosis, thrombosis, and vascular biology (2017-01-28)
Jean-Claude Tardif, David Rhainds, Eric Rhéaume, Marie-Pierre Dubé
ABSTRACT
High-density lipoproteins are involved in reverse cholesterol transport and possess anti-inflammatory and antioxidative properties. Paradoxically, CETP (cholesteryl ester transfer protein) inhibitors have been shown to increase inflammation as revealed by a raised plasma level of high-sensitivity C-reactive protein. CETP inhibitors did not improve clinical outcomes in large-scale clinical trials of unselected patients with coronary disease. Dalcetrapib is a CETP modulator for which effects on cardiovascular outcomes were demonstrated in the dal-OUTCOMES trial to be influenced by correlated polymorphisms in the
Sigma-Aldrich
Dalcetrapib, ≥97% (HPLC)