MilliporeSigma
  • Ceric Cyclopentadienides Bearing Alkoxy, Aryloxy, Chlorido, or Iodido Co-Ligands.
Ceric Cyclopentadienides Bearing Alkoxy, Aryloxy, Chlorido, or Iodido Co-Ligands.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-03-24)
David Schneider, Nicole Harmgarth, Frank T Edelmann, Reiner Anwander
ABSTRACT
A series of heteroleptic tris(cyclopentadienyl) Ce
Sigma-Aldrich
Diphenylphosphine, 98%
Sigma-Aldrich
Indene, ≥99%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%