MilliporeSigma
  • pH-Dependent Membrane Interactions of the Histidine-Rich Cell-Penetrating Peptide LAH4-L1.
pH-Dependent Membrane Interactions of the Histidine-Rich Cell-Penetrating Peptide LAH4-L1.
Biophysical journal (2017-07-25)
Justine Wolf, Christopher Aisenbrey, Nicole Harmouche, Jesus Raya, Philippe Bertani, Natalia Voievoda, Regine Süss, Burkhard Bechinger
ABSTRACT
The histidine-rich designer peptide LAH4-L1 exhibits antimicrobial and potent cell-penetrating activities for a wide variety of cargo including nucleic acids, polypeptides, adeno-associated viruses, and nanodots. The non-covalent complexes formed between the peptide and cargo enter the cell via an endosomal pathway where the pH changes from neutral to acidic. Here, we investigated the membrane interactions of the peptide with phospholipid bilayers and its membrane topology using static solid-state NMR spectroscopy. Oriented
Sigma-Aldrich
1,2-Dimyristoyl-d54-sn-glycero-3-phosphocholine, 98 atom % D, 97% (CP)
Sigma-Aldrich
Allyl methyl sulfone, 96%